missing translation for 'onlineSavingsMsg'
Learn More

5-Aminopyrimidine 98.0+%, TCI America™

Catalog No. A25891G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25891G Supplier TCI America Supplier No. A25891G
Only null left

Chemical Identifiers

CAS 591-55-9
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.105
MDL Number MFCD01529870
InChI Key FVLAYJRLBLHIPV-UHFFFAOYSA-N
PubChem CID 344373
ChEBI CHEBI:38619
IUPAC Name pyrimidin-5-amine
SMILES C1=C(C=NC=N1)N

Specifications

Melting Point 171°C
Color White
Quantity 1 g
Formula Weight 95.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminopyrimidine