missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-o-cresol 98.0+%, TCI America™

Catalog No. A0925500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0925500G 500 g
A092525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0925500G Supplier TCI America Supplier No. A0925500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2835-95-2
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00043922
InChI Key DBFYESDCPWWCHN-UHFFFAOYSA-N
Synonym 5-amino-o-cresol, 4-amino-2-hydroxytoluene, 3-hydroxy-4-methylaniline, phenol, 5-amino-2-methyl, 3-amino-6-methylphenol, 6-methyl-3-aminophenol, unii-ubb8xeb10b, 5-amino-2-methyl-phenol, 5-amino-2-methyl phenol, ccris 4582
PubChem CID 17818
IUPAC Name 5-amino-2-methylphenol
SMILES CC1=C(C=C(C=C1)N)O

Specifications

Melting Point 160°C
Quantity 500 g
Formula Weight 123.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-o-cresol
EINECSNumber (3)-0707&(9)-2068
RTECSNumber SJ6090000
TSCA Yes