missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-(4-fluorophenyl)-1H-pyrazole, 97%

Catalog No. AAH3283003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3283003 1 g
AAH3283006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3283003 Supplier Thermo Scientific Chemicals Supplier No. H3283003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 72411-52-0 | C9H8FN3 | 177.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72411-52-0
Molecular Formula C9H8FN3
Molecular Weight (g/mol) 177.18
MDL Number MFCD01023677
InChI Key QYEHDCXFXONDPV-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl-1h-pyrazol-5-amine, 5-4-fluorophenyl-1h-pyrazol-3-amine, 5-4-fluorophenyl-2h-pyrazol-3-ylamine, 5-amino-3-4-fluorophenyl-1h-pyrazole, 5-amino-3-4-fluorophenyl pyrazole, 1h-pyrazol-3-amine,5-4-fluorophenyl, 5-4-fluorophenyl-2h-pyrazol-3-amine, 3-amino-5-4-fluorophenyl-1h-pyrazole, 5-4-fluoro-phenyl-2h-pyrazol-3-ylamine, 3-4-fluorophenyl pyrazole-5-ylamine
PubChem CID 2759138
IUPAC Name 5-(4-fluorophenyl)-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1)C1=CC=C(F)C=C1

Specifications

Melting Point 122°C to 125°C
Quantity 1 g
Formula Weight 177.18
Percent Purity 97%
Chemical Name or Material 5-Amino-3-(4-fluorophenyl)-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only