missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3,4-dimethylisoxazole, 99%

Catalog No. AAL1866806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1866806 5 g
AAL1866803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1866806 Supplier Thermo Scientific Chemicals Supplier No. L1866806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19947-75-2 | C5H8N2O | 112.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19947-75-2
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD00046081
InChI Key PYNDWPFZDQONDV-UHFFFAOYSA-N
Synonym 5-amino-3,4-dimethylisoxazole, 3,4-dimethylisoxazol-5-amine, 5-amino-3,4-dimethyl-isoxazole, 5-isoxazolamine, 3,4-dimethyl, dimethyl-1,2-oxazol-5-amine, 3,4-dimethyl-5-isoxazolamine, 5-amino-3,4-dimethyl isoxazole, 3,4-dimethylisoxazole-5-ylamine, 3,4-dimethylisoxazol-5-ylamine, pubchem8660
PubChem CID 88317
IUPAC Name 3,4-dimethyl-1,2-oxazol-5-amine
SMILES CC1=C(ON=C1C)N

Specifications

Melting Point 118°C to 120°C
Quantity 5 g
Beilstein 108546
Formula Weight 112.13
Percent Purity 99%
Chemical Name or Material 5-Amino-3,4-dimethylisoxazole
EINECSNumber 243-437-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only