missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylbenzenesulfonamide 98.0+%, TCI America™

Catalog No. A25525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25525G 5 g
A255225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25525G Supplier TCI America Supplier No. A25525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6973-09-7
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD06681071
InChI Key KTPBKMYOIFHJMI-UHFFFAOYSA-N
Synonym 3-amino-6-methylbenzenesulfonamide, 2-methyl-5-aminobenzene sulfonamide, 5-amino-2-methylbenzene-1-sulfonamide, benzenesulfonamide, 5-amino-2-methyl, 5-amino-2-methyl-benzenesulfonamide, 3-aminosulfonyl-4-methylaniline, pubchem14390, p-aminotoluol-sulfonamid, acmc-1bcyh, ksc495k6b
PubChem CID 235511
IUPAC Name 5-amino-2-methylbenzenesulfonamide
SMILES CC1=C(C=C(C=C1)N)S(=O)(=O)N

Specifications

Melting Point 169°C
Quantity 5 g
Formula Weight 186.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-methylbenzenesulfonamide