missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methoxyphenol, 98%

Catalog No. AAL1023414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1023414 25 g
AAL1023406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1023414 Supplier Thermo Scientific Chemicals Supplier No. L1023414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1687-53-2 | C7H9NO2 | 139.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1687-53-2
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00010222
InChI Key BLQFHJKRTDIZLX-UHFFFAOYSA-N
Synonym 3-hydroxy-4-methoxyaniline, 5-aminoguaiacol, 2-methoxy-5-aminophenol, phenol, 5-amino-2-methoxy, 5-amino-2-methoxy-phenol, pubchem3112, acmc-209dyt, 3-hydroxy4-methoxyaniline, 5-amino-2-methoxy phenol, 3-hydroxy-4-methoxylaniline
PubChem CID 74314
IUPAC Name 5-amino-2-methoxyphenol
SMILES COC1=C(C=C(C=C1)N)O

Specifications

Melting Point 130°C to 132°C
Quantity 25 g
Beilstein 2717084
Formula Weight 139.16
Percent Purity 98%
Chemical Name or Material 5-Amino-2-methoxyphenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-873-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only