missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methoxybenzoic Acid 97.0+%, TCI America™

Catalog No. A21841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21841G Supplier TCI America Supplier No. A21841G

Chemical Identifiers

CAS 3403-47-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD09260887
InChI Key LWWPSEIFAKNPKQ-UHFFFAOYSA-N
PubChem CID 13549244
IUPAC Name 5-amino-2-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)N)C(=O)O

Specifications

Melting Point 165°C
Color Red-Yellow
Quantity 1 g
Formula Weight 167.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-methoxybenzoic Acid