missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-fluoropyridine 98.0+%, TCI America™

Catalog No. A23841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23841G Supplier TCI America Supplier No. A23841G
Only null left

Chemical Identifiers

CAS 1827-27-6
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.11
MDL Number MFCD01632180
InChI Key YTHMOBMZVVFNBE-UHFFFAOYSA-N
Synonym 5-amino-2-fluoropyridine, 2-fluoro-5-aminopyridine, 3-amino-6-fluoropyridine, 6-fluoro-pyridin-3-ylamine, 6-fluoro-3-pyridinamine, 3-pyridinamine, 6-fluoro, 6-fluoro-3-pyridylamine, 6-fluoro-3-pyridineamine, 5-amine-2-fluoropyridine, 6-fluoropyridine-3-amine
PubChem CID 819440
IUPAC Name 6-fluoropyridin-3-amine
SMILES NC1=CN=C(F)C=C1

Specifications

Melting Point 90°C
Color Purple
Quantity 1 g
Formula Weight 112.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-fluoropyridine