missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-fluoropyridine, 97+%, Thermo Scientific Chemicals

Catalog No. AAH3087206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3087206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3087206 Supplier Thermo Scientific Chemicals Supplier No. H3087206
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1827-27-6
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.11
MDL Number MFCD01632180
InChI Key YTHMOBMZVVFNBE-UHFFFAOYSA-N
Synonym 5-amino-2-fluoropyridine, 2-fluoro-5-aminopyridine, 3-amino-6-fluoropyridine, 6-fluoro-pyridin-3-ylamine, 6-fluoro-3-pyridinamine, 3-pyridinamine, 6-fluoro, 6-fluoro-3-pyridylamine, 6-fluoro-3-pyridineamine, 5-amine-2-fluoropyridine, 6-fluoropyridine-3-amine
PubChem CID 819440
IUPAC Name 6-fluoropyridin-3-amine
SMILES NC1=CN=C(F)C=C1

Specifications

Melting Point 88°C to 91°C
Odor Characteristic
Quantity 5 g
UN Number UN2811
Formula Weight 112.11
Percent Purity ≥97%
Chemical Name or Material 5-Amino-2-fluoropyridine
Hazard Category H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No

RUO – Research Use Only