missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-bromobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7117603
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18475G 5 g
A184725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18475G Supplier TCI America Supplier No. A18475G

Chemical Identifiers

CAS 393-36-2
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
MDL Number MFCD00007827
InChI Key YGNISOAUPSJDJE-UHFFFAOYSA-N
Synonym 4-bromo-3-trifluoromethyl aniline, 5-amino-2-bromobenzotrifluoride, 4-bromo-3-trifluoromethyl-phenylamine, 3-trifluoromethyl-4-bromoaniline, 4-amino-1-bromo-2-trifluoromethyl benzene, 4-bromo-alpha,alpha,alpha-trifluoro-m-toluidine, benzenamine, 4-bromo-3-trifluoromethyl, 5-amino-2-bromo benzotrifluoride, buttpark 37\11-68
PubChem CID 67852
IUPAC Name 4-bromo-3-(trifluoromethyl)aniline
SMILES NC1=CC=C(Br)C(=C1)C(F)(F)F

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 108°C
Quantity 5 g
Formula Weight 240.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-2-bromobenzotrifluoride