Learn More
5-Amino-2-bromo-3-fluoropyridine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H3315403
| Quantity | 1g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsCommercially available 5-bromo-2-fluoropyridine was treated with LDA at -78°C to give the lithiated species. Fluoropyridine-based bromoacetamide reagent has been prepared and used in prosthetic group introduction for the labeling of oligonucleotides.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate(2-7°C ).
Chemical Identifiers
| 1256276-41-1 | |
| 191.003 | |
| DDJLNVAJTGIWNJ-UHFFFAOYSA-N | |
| 71741382 | |
| C1=C(C=NC(=C1F)Br)N |
| C5H4BrFN2 | |
| MFCD21099397 | |
| 5-amino-2-bromo-3-fluoropyridine, 3-pyridinamine, 6-bromo-5-fluoro | |
| 6-bromo-5-fluoropyridin-3-amine |
Specifications
| 5-Amino-2-bromo-3-fluoropyridine | |
| C5H4BrFN2 | |
| MFCD21099397 | |
| DDJLNVAJTGIWNJ-UHFFFAOYSA-N | |
| 6-bromo-5-fluoropyridin-3-amine | |
| 71741382 | |
| 97% |
| 1256276-41-1 | |
| 1g | |
| 5-amino-2-bromo-3-fluoropyridine, 3-pyridinamine, 6-bromo-5-fluoro | |
| C1=C(C=NC(=C1F)Br)N | |
| 191.003 | |
| 191 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures