missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,4-dichloropyrimidine, 97%, Thermo Scientific Chemicals

Catalog No. AAH3260606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3260606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3260606 Supplier Thermo Scientific Chemicals Supplier No. H3260606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 5177-27-5 | Molecular Formula: C4H3Cl2N3 | Molecular Weight: 163.989 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5177-27-5
Molecular Formula C4H3Cl2N3
Molecular Weight (g/mol) 163.989
MDL Number MFCD05662684
InChI Key RINHVELYMZLXIW-UHFFFAOYSA-N
PubChem CID 257797
IUPAC Name 2,4-dichloropyrimidin-5-amine
SMILES C1=C(C(=NC(=N1)Cl)Cl)N

Specifications

Quantity 5 g
Synonym 5-amino-2,4-dichloropyrimidine, 2,4-dichloro-5-aminopyrimidine, 2,4-dichloro-pyrimidin-5-ylamine, 2,4-dichloro-5-pyrimidinamine, 5-amino-2, 4-dichloropyrimidine, 5-pyrimidinamine, 2,4-dichloro, 2,4-dichlorpyrimidin-5-amin, pubchem21459, acmc-209kvs, ksc497e8n
Formula Weight 164
Percent Purity 97%
Chemical Name or Material 5-Amino-2,4-dichloropyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only