missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,3-dihydrobenzofuran, 97%

Catalog No. AC438030050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438030050 5 g
AC438030010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438030050 Supplier Thermo Scientific Chemicals Supplier No. 438030050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42933-43-7 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42933-43-7
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
InChI Key YJMADHMYUJFMQE-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-amine, 2,3-dihydrobenzo b furan-5-ylamine, 5-amino-2,3-dihydrobenzo b furan, 5-aminocoumaran, 5-amino-2,3-dihydrobenzofuran, 2,3-dihydro-5-benzofuranamine, 2,3-dihydrobenzo b furan-5-amine, pubchem16462, 2,3-dihydro-5-benzofuranamine;, 2,3-dihydrobenzofuran-5-amine;
PubChem CID 3841102
IUPAC Name 2,3-dihydro-1-benzofuran-5-amine
SMILES C1COC2=C1C=C(C=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 81.6°C to 83.4°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 135.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Amino-2, 3-dihydrobenzofuran
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Co

RUO – Research Use Only