missing translation for 'onlineSavingsMsg'
Learn More

5-Allyl-3-methoxysalicylaldehyde 98.0+%, TCI America™

Catalog No. A21045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21045G 5 g
A21041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21045G Supplier TCI America Supplier No. A21045G

Chemical Identifiers

CAS 22934-51-6
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD06408022
InChI Key FDHXEIOBIOVBEN-UHFFFAOYSA-N
Synonym 5-allyl-2-hydroxy-3-methoxybenzaldehyde, 5-allyl-3-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-3-methoxy-5-2-propenyl, benzaldehyde,2-hydroxy-3-methoxy-5-2-propen-1-yl, 2-hydroxy-3-methoxy-5-prop-2-en-1-yl benzaldehyde, formyl eugenol, acmc-209fzq, 2-hydroxy-3-methoxy-5-2-propenyl benzaldehyde, 3-methoxy-2-oxidanyl-5-prop-2-enyl-benzaldehyde
PubChem CID 1713887
IUPAC Name 2-hydroxy-3-methoxy-5-prop-2-enylbenzaldehyde
SMILES COC1=C(C(=CC(=C1)CC=C)C=O)O

Specifications

Melting Point 48°C
Color Yellow
Boiling Point 167°C
Quantity 5 g
Formula Weight 192.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Allyl-3-methoxysalicylaldehyde