missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dihydroxyflavone, 98%

Catalog No. AAL1417814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1417814 25 g
AAL1417806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1417814 Supplier Thermo Scientific Chemicals Supplier No. L1417814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 480-40-0 | C15H10O4 | 254.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 480-40-0
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00006834
InChI Key RTIXKCRFFJGDFG-UHFFFAOYSA-N
Synonym chrysin, 5,7-dihydroxyflavone, chrysine, 5,7-dihydroxy-2-phenyl-4h-chromen-4-one, crysin, 4h-1-benzopyran-4-one, 5,7-dihydroxy-2-phenyl, unii-3cn01f5zj5, flavone, 5,7-dihydroxy, 5,7-dihydroxy-2-phenyl-4h-1-benzopyran-4-one, chembl117
PubChem CID 5281607
ChEBI CHEBI:75095
IUPAC Name 5,7-dihydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O

Specifications

Melting Point 284°C to 289°C
Quantity 25 g
Beilstein 233276
Merck Index 14,2256
Formula Weight 254.24
Percent Purity 98%
Chemical Name or Material 5,7-Dihydroxyflavone
EINECSNumber 207-549-7
RTECSNumber LK8329050
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only