missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichloropyrazolo[1,5-a]pyrimidine, 95%, Thermo Scientific Chemicals

Catalog No. AC454130010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454130010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454130010 Supplier Thermo Scientific Chemicals Supplier No. 454130010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57489-77-7
Molecular Formula C6H3Cl2N3
Molecular Weight (g/mol) 188.02
InChI Key JMTFWCYVZOFHLR-UHFFFAOYSA-N
Synonym 5,7-dichloropyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 5,7-dichloro, 5,7-dichloro-pyrazolo 1,5-a pyrimidine, pubchem14638, 5,7-dichloropyrazolo 1,5-a pyri-midine, 5, 7-dichloro-pyrazolo 1,5-a pyrimidine, 5,7-bis chloranyl pyrazolo 1,5-a pyrimidine
PubChem CID 11074154
IUPAC Name 5,7-dichloropyrazolo[1,5-a]pyrimidine
SMILES C1=C2N=C(C=C(N2N=C1)Cl)Cl

Specifications

Melting Point 75.0°C to 76.5°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 188.02
Percent Purity 95%
Chemical Name or Material 5,7-Dichloropyrazolo[1,5-a]pyrimidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only