missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dibromo-8-hydroxyquinoline 98.0+%, TCI America™

Catalog No. D020825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D020825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D020825G Supplier TCI America Supplier No. D020825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 5,7-Dibromo-8-hydroxyquinoline
CAS 521-74-4
Melting Point 199°C
Molecular Formula C9H5Br2NO
Quantity 25 g
MDL Number MFCD00006785
Synonym broxyquinoline, 5,7-dibromo-8-hydroxyquinoline, 5,7-dibromo-8-quinolinol, broxyquinolin, dibromoxin, dibromoxine, dibromoxyquinoline, broxiquinoline, dibromoquin, broxykinolin
InChI Key ZDASUJMDVPTNTF-UHFFFAOYSA-N
SMILES C1=CC2=C(C(=C(C=C2Br)Br)O)N=C1
IUPAC Name 5,7-dibromoquinolin-8-ol
Molecular Weight (g/mol) 302.953
PubChem CID 2453
Formula Weight 302.95
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Show More Show Less