missing translation for 'onlineSavingsMsg'
Learn More

5(6)-Tetramethylrhodamine isothiocyanate

Numéro de catalogue. AAJ62258MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAJ62258MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AAJ62258MA Fournisseur Thermo Scientific Chemicals Code fournisseur J62258MA
Il en reste null

CAS: 95197-95-8 | C25H21N3O3S

5(6)-TRITC is widely used in preparing bioconjugates of proteins and nucleic acids. It is suitable for protein, peptide and nucleotide labeling. Serves as an acceptor for FRET. It is recommended as a donor molecule in these FRET-based applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5(6)-TRITC is widely used in preparing bioconjugates of proteins and nucleic acids. It is suitable for protein, peptide and nucleotide labeling. Serves as an acceptor for FRET. It is recommended as a donor molecule in these FRET-bas

Identifiants chimiques

CAS 95197-95-8
Molecular Formula C25H21N3O3S
MDL Number MFCD00063517
Synonym TRITC; MRITC

Spécifications

Color Purple to Red
Quantity 10 mg
Sensitivity Light, Air and Moisture Sensitive
Solubility Information Soluble in DMSO or methanol
Formula Weight 443.52
Physical Form Powder
Chemical Name or Material 5(6)-Tetramethylrhodamine isothiocyanate
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H334-H335-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only