missing translation for 'onlineSavingsMsg'
Learn More

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4in.-disulfonic acid monosodium salt hydrate, 97%

Catalog No. AAB2406614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2406614 25 g
AAB2406603 1 g
AAB2406606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2406614 Supplier Thermo Scientific Chemicals Supplier No. B2406614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1266615-85-3 | C20H15N4NaO7S2 | 510.471 g/mol

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4-disulfonic acid monosodium salt hydrate is used in the accurate determination of sub microgram levels of serum iron. (i.e.. reagent for the spectrophotometric determination of Fe).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4″-disulfonic acid monosodium salt hydrate is used in the accurate determination of sub microgram levels of serum iron. (i.e.. reagent for the spectrophotometric determination of Fe).

Solubility
Soluble in water (partly).

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1266615-85-3
Molecular Formula C20H15N4NaO7S2
Molecular Weight (g/mol) 510.471
MDL Number MFCD00150794
InChI Key MBOKSYFCYXIJRZ-UHFFFAOYSA-M
Synonym ferrozine mono-sodium salt hydrate, pdt disulfonate monosodium salt hydrate, 5,6-diphenyl-3-2-pyridyl-1,2,4-triazine-4,4-disulfonic acid monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p inverted exclamation marka-disulfonic acid monosodium salt hydrate, 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid monosodium salt hydrate, sodium 4-3-pyridin-2-yl-6-4-sulfophenyl-1,2,4-triazin-5-yl benzenesulfonate hydrate
PubChem CID 71311205
IUPAC Name sodium;4-[3-pyridin-2-yl-6-(4-sulfophenyl)-1,2,4-triazin-5-yl]benzenesulfonate;hydrate
SMILES C1=CC=NC(=C1)C2=NC(=C(N=N2)C3=CC=C(C=C3)S(=O)(=O)O)C4=CC=C(C=C4)S(=O)(=O)[O-].O.[Na+]

Specifications

Quantity 25 g
Beilstein 5222087
Solubility Information Soluble in water (partly).
Formula Weight 492.47 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulfonic acid monosodium salt hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 274-196-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only