missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro-2H-pyran-2-one 95.0+%, TCI America™

Catalog No. D22615G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22615G 5 g
D22611G 1 g
D226125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D22615G Supplier TCI America Supplier No. D22615G

Chemical Identifiers

CAS 3393-45-1
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00010439
InChI Key VREFDQNWPNDZEX-UHFFFAOYSA-N
Synonym 5,6-dihydro-2h-pyran-2-one, 2h-pyran-2-one, 5,6-dihydro, 5,6-dihydropyran-2-one, 5,6-dihydro-pyran-2-one, chembl55078, 5,6-dihydro-2-pyranone, acmc-1ck70, 2-pentenoic acid, 5-hydroxy-, .delta.-lactone, 5,6-dihydro-2h-pyran-2-one, technical grade
PubChem CID 520660
IUPAC Name 3,6-dihydro-2H-pyran-2-one
SMILES O=C1CC=CCO1

Specifications

Color Yellow
Boiling Point 117°C
Quantity 5 g
UN Number 1224
Formula Weight 98.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5,6-Dihydro-2H-pyran-2-one