missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dihydro-1,4-dithiin-2,3-dicarboximide 97.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 217°C |
| Quantity | 1 g |
| Formula Weight | 187.23 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5,6-Dihydro-1,4-dithiin-2,3-dicarboximide |
Sécurité et manipulation
| RTECSNumber | JO6955000 |
| TSCA | No |