missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Indigodisulfonic Acid, Disodium Salt, BAKER™, J.T. Baker™

Catalog No. 02002110
Change view
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002110 100 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02002110 Supplier Avantor Supplier No. N87705

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 860-22-0
Molecular Formula C16H8N2Na2O8S2
Molecular Weight (g/mol) 466.35
MDL Number MFCD00005723
InChI Key KHLVKKOJDHCJMG-QDBORUFSSA-L
Synonym indigo carmine, indigocarmine, acid blue 74, indigocarmin, indigotindisulfonate sodium, c.i. acid blue 74, food blue no. 2, amacid brilliant blue, c.i. food blue 1, fd&c blue no. 2
PubChem CID 5284351
IUPAC Name disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 100 g
Formula Weight 466.36
Chemical Name or Material 5,5'-Indigodisulfonic Acid, Disodium Salt