missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ 5,5'-Dithio-bis-(2-nitrobenzoic Acid), Calbiochem™,

Catalog No. 3221235GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3221235GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3221235GM Supplier MilliporeSigma™ Supplier No. 3221235GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfhydryl reagent used to characterize reactive SH groups

Chemical Identifiers

CAS 69-78-3
Molecular Formula C14H8N2O8S2
Molecular Weight (g/mol) 396.34
MDL Number MFCD00007140
InChI Key KIUMMUBSPKGMOY-UHFFFAOYSA-N
Synonym dtnb, 5,5'-dithiobis 2-nitrobenzoic acid, ellman's reagent, 3-carboxy-4-nitrophenyl disulfide, dithionitrobenzoic acid, dithiobisnitrobenzoic acid, 5,5'-disulfanediylbis 2-nitrobenzoic acid, benzoic acid, 3,3'-dithiobis 6-nitro, 3,3'-dithiobis 6-nitrobenzoic acid, 5,5'-dithio-bis 2-nitrobenzoic acid
PubChem CID 6254
ChEBI CHEBI:86228
IUPAC Name 5-[(3-carboxy-4-nitrophenyl)disulfanyl]-2-nitrobenzoic acid
SMILES OC(=O)C1=CC(SSC2=CC=C(C(=C2)C(O)=O)[N+]([O-])=O)=CC=C1[N+]([O-])=O

Specifications

Color Yellow
Quantity 5 g
Solubility Information Soluble: EtOH
Formula Weight 396.4
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 5,5'-Dithio-bis-(2-nitrobenzoic Acid)
RTECSNumber DG9650000
Recommended Storage 2° to 8°C (35.6° to 46.4°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 4°C (39.2°F).