missing translation for 'onlineSavingsMsg'
Learn More

5,5-Diphenylhydantoin, 99%

Catalog No. AAA1701314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1701314 25 g
AAA1701322 100 g
AAA1701336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1701314 Supplier Thermo Scientific Chemicals Supplier No. A1701314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57-41-0 | C15H12N2O2 | 252.273 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-41-0
Molecular Formula C15H12N2O2
Molecular Weight (g/mol) 252.273
MDL Number MFCD00005264
InChI Key CXOFVDLJLONNDW-UHFFFAOYSA-N
Synonym phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan
PubChem CID 1775
ChEBI CHEBI:8107
IUPAC Name 5,5-diphenylimidazolidine-2,4-dione
SMILES C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3

Specifications

Melting Point 293°C to 298°C
Quantity 25 g
Beilstein 384532
Merck Index 14,7322
Formula Weight 252.27
Percent Purity 99%
Chemical Name or Material 5,5-Diphenylhydantoin
Hazard Category H302-H350-H361d-H500
Hazard Statement GHS H Statement
H350-H360-H317
May cause cancer.
May damage fertility or the unborn child.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 200-328-6
RTECSNumber MU1050000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only