missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dimethyl-1-pyrroline N-Oxide 97.0+%, TCI America™

Catalog No. D23625G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23625G 5 g
D23621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23625G Supplier TCI America Supplier No. D23625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3317-61-1
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00005279
InChI Key VCUVETGKTILCLC-UHFFFAOYSA-N
Synonym 5,5-dimethyl-1-pyrroline n-oxide, dmpo, 5,5-dimethyl-1-pyrroline-n-oxide, 5,5-dimethyl-1-pyrroline-1-oxide, 2,2-dimethyl-3,4-dihydro-2h-pyrrole 1-oxide, 2,2-dimethyl-3,4-dihydropyrrol-1-ium-1-olate, 2h-pyrrole, 3,4-dihydro-2,2-dimethyl-, 1-oxide, unii-7170jz1qf3, acmc-209hyz, lopac-d-5766
PubChem CID 1774
IUPAC Name 2,2-dimethyl-1-oxido-3,4-dihydropyrrol-1-ium
SMILES CC1(CCC=[N+]1[O-])C

Specifications

Quantity 5 g
Formula Weight 113.16
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5,5-Dimethyl-1-pyrroline N-Oxide
TSCA No
Recommended Storage Freezer