missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dimethyl-1-pyrroline N-oxide, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013943
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013943 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013943 Supplier Merck Emd Millipore Supplier No. 92688100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00005279
Synonym DMPO

Specifications

Melting Point 25°C to 29°C
CAS 3317-61-1
Density 1.015 g/mL (at 25°C)
Boiling Point 75°C/0.4 mmHg
Flash Point 95°C
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C6H11 NO
Quantity 100 mg
Formula Weight 113.16
Percent Purity ≥98.0% (GC)
Grade For ESR-spectroscopy
Show More Show Less