missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dimethyl-1,3-dioxan-2-one, tech.

Catalog No. AAL0876703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0876703 1 g
AAL0876706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0876703 Supplier Thermo Scientific Chemicals Supplier No. L0876703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3592-12-9 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3592-12-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00014650
InChI Key JRFXQKZEGILCCO-UHFFFAOYSA-N
Synonym neopentylene carbonate, 2,2-dimethyltrimethylene carbonate, 5,5-dimethyl-1,3-diox-2-one, acmc-1adx1, 3-nitro-5-chlorobenzenesulfonamide, 5,5-dimethyltrimethylene carbonate, 5,5-dimethyl-1,3-dioxane-2-one, 5,5-dimethyl-2-oxo-1,3-dioxane, 1,3-dioxan-2-one,5,5-dimethyl, 5,5-dimethyl-1,3 dioxan-2-one
PubChem CID 137984
IUPAC Name 5,5-dimethyl-1,3-dioxan-2-one
SMILES CC1(COC(=O)OC1)C

Specifications

Melting Point 105°C to 114°C
Quantity 1 g
Beilstein 109055
Formula Weight 130.14
Grade Technical
Chemical Name or Material 5,5-Dimethyl-1,3-dioxan-2-one
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only