missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Methylphenyl)-1H-tetrazole, 99%

Catalog No. AAL19263MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19263MD 250 mg
AAL1926303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19263MD Supplier Thermo Scientific Chemicals Supplier No. L19263MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3441-00-7 | C8H8N4 | 160.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3441-00-7
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD03093096
InChI Key GWZNHKMUSLKULV-UHFFFAOYSA-N
Synonym 5-3-methylphenyl-1h-tetrazole, 5-m-tolyl-1h-tetrazole, 5-3-methylphenyl tetrazole, 2h-tetrazole,5-3-methylphenyl, 5-3-methylphenyl-1h-1,2,3,4-tetrazole, 3-tolyl tetrazole, 5-m-tolyl tetrazole, acmc-1cmun, 5-m-tolyl 1h tetrazole
PubChem CID 583308
IUPAC Name 5-(3-methylphenyl)-2H-tetrazole
SMILES CC1=CC=CC(=C1)C2=NNN=N2

Specifications

Melting Point 143°C to 146°C
Quantity 250 mg
Beilstein 131205
Formula Weight 160.18
Percent Purity 99%
Chemical Name or Material 5-(3-Methylphenyl)-1H-tetrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only