missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Thienyl)pentanoic acid, 95%

Catalog No. AAL1312206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1312206 5 g
AAL1312203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1312206 Supplier Thermo Scientific Chemicals Supplier No. L1312206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21010-06-0 | C9H12O2S | 184.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21010-06-0
Molecular Formula C9H12O2S
Molecular Weight (g/mol) 184.25
MDL Number MFCD00151849
InChI Key OQFTZRHAQGXEQU-UHFFFAOYSA-N
Synonym 5-2-thienyl pentanoic acid, 5-thiophen-2-yl pentanoic acid, 2-thiophenepentanoic acid, 5-2-thienyl valeric acid, maybridge3_003072, acmc-1cq6n, 5-2-thienyl-pentanoic acid, 5-2-thienyl-n-valeric acid
PubChem CID 152324
IUPAC Name 5-thiophen-2-ylpentanoic acid
SMILES OC(=O)CCCCC1=CC=CS1

Specifications

Melting Point 39°C to 42°C
Quantity 5 g
Beilstein 126021
Formula Weight 184.26
Percent Purity 95%
Chemical Name or Material 5-(2-Thienyl)pentanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only