missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Furyl)-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Numéro de catalogue. p-3110734
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC35001DA 1g
CC35001DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC35001DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC35001DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 116153-81-2
Formule moléculaire C8H6N2O3
Poids moléculaire (g/mol) 178.147
Numéro MDL MFCD05170057
Clé InChI GKPSFQIKCROJOB-UHFFFAOYSA-N
Synonyme 5-2-furyl-1h-pyrazole-3-carboxylic acid, 5-furan-2-yl-1h-pyrazole-3-carboxylic acid, 5-furan-2-yl-2h-pyrazole-3-carboxylic acid, 3-furan-2-yl-1h-pyrazole-5-carboxylic acid, 3-2-furyl-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-2-furanyl, 5-2-furyl pyrazole-3-carboxylic acid, acmc-20f1sn, 3-2-furyl pyrazole-5-carboxylic acid
PubChem CID 654018
Nom de l’IUPAC 5-(furan-2-yl)-1H-pyrazole-3-carboxylic acid
SOURIRES C1=COC(=C1)C2=CC(=NN2)C(=O)O

Spécifications

Nom ou substance chimique 5-(2-Furyl)-1H-pyrazole-3-carboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 1g
Poids de formule 178.15
Pourcentage de pureté 97%
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.