missing translation for 'onlineSavingsMsg'
Learn More

5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin 95.0+%, TCI America™

Catalog No. T1730100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1730100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1730100MG Supplier TCI America Supplier No. T1730100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25440-14-6
Molecular Formula C44H10F20N4
Molecular Weight (g/mol) 974.56
MDL Number MFCD00010032
InChI Key GBAPBSUXEFFUAD-UHFFFAOYSA-N
Synonym 5,10,15,20-tetrakis pentafluorophenyl porphyrin, 5,10,15,20-tetrakis pentafluorophenyl-21h,23h-porphine, meso-tetra pentafluorophenyl porphine, 21h,23h-porphine, 5,10,15,20-tetrakis pentafluorophenyl, 5,10,15,20-tetrakis 2,3,4,5,6-pentafluorophenyl-21,22-dihydroporphyrin, 2,7,12,17-tetrakis 2,3,4,5,6-pentafluorophenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaene, acmc-209gjw, iron iii ionophore iv, tetra pentafluorophenyl poephine, tetrakis pentafluorophenyl porphyrin
PubChem CID 3671775
IUPAC Name 2,7,12,17-tetrakis(2,3,4,5,6-pentafluorophenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
SMILES FC1=C(F)C(F)=C(C(F)=C1F)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Color Black-Purple
Quantity 100 mg
Formula Weight 974.56
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin