missing translation for 'onlineSavingsMsg'
Learn More

5-(1,4-Diazepan-1-yl)-3-phenyl-1,2,4-thiadiazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals AW00426CB

Quantity 250mg

 View more versions of this product

Catalog No. AW00426CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

306934-71-4
260.36
IENGAZSTLAONAR-UHFFFAOYSA-N
2737169
C1CNCCN(C1)C1=NC(=NS1)C1=CC=CC=C1
C13H16N4S
MFCD00729015
5-1,4-diazepan-1-yl-3-phenyl-1,2,4-thiadiazole, 1-3-phenyl-1,2,4-thiadiazol-5-yl-1,4-diazepane, 1h-1,4-diazepine,hexahydro-1-3-phenyl-1,2,4-thiadiazol-5-yl, maybridge1_000443, 1-3-phenyl-1,2,4-thiazol-5-yl homopiperazine, 5-1,4-diazaperhydroepinyl-3-phenyl-1,2,4-thiadiazole
5-(1,4-diazepan-1-yl)-3-phenyl-1,2,4-thiadiazole

Specifications

5-(1,4-Diazepan-1-yl)-3-phenyl-1,2,4-thiadiazole
96.5
97%
C13H16N4S
MFCD00729015
IENGAZSTLAONAR-UHFFFAOYSA-N
5-(1,4-diazepan-1-yl)-3-phenyl-1,2,4-thiadiazole
2737169
97%
306934-71-4
100.0
Amber glass bottle
250mg
5-1,4-diazepan-1-yl-3-phenyl-1,2,4-thiadiazole, 1-3-phenyl-1,2,4-thiadiazol-5-yl-1,4-diazepane, 1h-1,4-diazepine,hexahydro-1-3-phenyl-1,2,4-thiadiazol-5-yl, maybridge1_000443, 1-3-phenyl-1,2,4-thiazol-5-yl homopiperazine, 5-1,4-diazaperhydroepinyl-3-phenyl-1,2,4-thiadiazole
C1CNCCN(C1)C1=NC(=NS1)C1=CC=CC=C1
260.36
260.36
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions