Learn More
Sklar Instruments Scissors, Merit Operating Scissors, Sklar 5-1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA.
Numéro de catalogue. 501094110
Encompass
Scissors, Merit Operating Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA. Merit Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.
Numéro de catalogue. 501094110 Fournisseur Sklar Instruments Code fournisseur 97299
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Scissors, Merit Operating Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA. Merit Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.