Learn More
Alkali Scientific 4mm Electroporation Cuvettes, Square Lid, Sterile, 50/cs
Numéro de catalogue. 502388902
Encompass
Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies. Precision built to give exacting tolerances ensuring reproducible results. Polished aluminum plates reduce static and arcing for optimum performance. Individually wrapped and gamma irradiated providing guaranteed sterility. Compatible with most Electroporation devices. Individually color coded cuvette caps identify gap widths of 4mm. Each bag contains 50 individually wrapped sterile cuvettes.
Numéro de catalogue. 502388902 Fournisseur Alkali Scientific Code fournisseur EP404004
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies. Precision built to give exacting tolerances ensuring reproducible results. Polished aluminum plates reduce static and arcing for optimum performance. Individually wrapped and gamma irradiated providing guaranteed sterility. Compatible with most Electroporation devices. Individually color coded cuvette caps identify gap widths of 4mm. Each bag contains 50 individually wrapped sterile cuvettes.