missing translation for 'onlineSavingsMsg'
Learn More

4H-Thieno[3,2-b]pyrrole-5-carboxylic acid, 97%, Thermo Scientific™

Numéro de catalogue. CC39001CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC39001CB 250mg
CC39001DA 1g
CC39001DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC39001CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC39001CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 39793-31-2
Formule moléculaire C7H5NO2S
Poids moléculaire (g/mol) 167.182
Numéro MDL MFCD07368542
Clé InChI PMHDSACGRKBACK-UHFFFAOYSA-N
Synonyme 4h-thieno 3,2-b pyrrole-5-carboxylic acid, 4h-thieno 3,2-b pyrole-5-carboxylic acid, 4wl, d-amino acid oxidase inhibitor, 4h-thieno 3,2-b pyrrole-5-carboxylicacid, thiopheno 3,2-b pyrrole-5-carboxylic acid, 4h-thieno 2,3-d pyrrole-5-carboxylic acid, 4h-thieno 3 pound not2-b pyrrole-5-carboxylic acid
PubChem CID 4961254
Nom de l’IUPAC 4H-thieno[3,2-b]pyrrole-5-carboxylic acid
SOURIRES C1=CSC2=C1NC(=C2)C(=O)O

Spécifications

Nom ou substance chimique 4H-Thieno[3, 2-b]pyrrole-5-carboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 250mg
Poids de formule 167.19
Pourcentage de pureté 97%
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.