missing translation for 'onlineSavingsMsg'
Learn More

4',6-Diamidino-2-Phenylindole Dihydrochloride MP Biomedicals

Catalog No. p-3178563
Change view
Click to view available options
Quantity:
1 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15757401 1 mg
ICN15757450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15757401 Supplier MP Biomedicals Inc Supplier No. 0215757401

Chemical Identifiers

CAS 28718-90-3
Molecular Formula C16H17Cl2N5
Molecular Weight (g/mol) 350.25
MDL Number MFCD00012681
InChI Key FPNZBYLXNYPRLR-UHFFFAOYSA-N
Synonym 4',6-diamidino-2-phenylindole dihydrochloride, dapi dihydrochloride, dapi, dihydrochloride, 2-4-carbamimidoylphenyl-1h-indole-6-carboximidamide dihydrochloride, dapi hydrochloride, 2-phenylindole-4',6-dicarboxamidine dihydrohydrochloride hydrate, 2-4-amidinophenyl-6-indolecarbamidine dihydrochloride, 4′,6-diamidino-2-phenylindole, 2hcl, ccris 8836
PubChem CID 160166
IUPAC Name dihydrogen 2-(4-carbamimidoylphenyl)-1H-indole-6-carboximidamide dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NC(=N)C1=CC=C(C=C1)C1=CC2=CC=C(C=C2N1)C(N)=N

Specifications

Quantity 1 mg
Formula Weight 350.2
Chemical Name or Material 4′, 6-Diamidino-2-Phenylindole Dihydrochloride