Learn More
Justrite™ Fire Protection Extinguisher Unit
Catalog No. 19414040
Change view
Click to view available options
For Use With (Equipment):
2-drum locker
4-,6-,9-,12-,16-drum lockers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19414040 2-drum locker
19414041 4-,6-,9-,12-,16-drum lockers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19414040 Supplier Justrite™ Supplier No. 915401
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Accessory for Outdoor Safety Lockers
  • For use with 2-,4-,6-,9-,12-,16-drum lockers only
  • Includes FE-227, BC type extinguisher unit
  • Ceiling mounted, fuse linked
  • Accessory must be purchased in conjunction with locker to ensure proper installation
Applications:
Adds fire protection system as recommended for storage of flammable or combustible materials
Specifications
Product Type BC Type Fire Extinguisher Unit
For Use With (Equipment) 2-drum locker
Includes FE-227, BC type extinguisher unit