missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Biohazard Identification Signs: Biological Hazard Authorized Personnel Only

For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents

$20.40 - $36.41

Specifications

Height (English) 10 in.
Height (Metric) 3 m
Width (English) 14 in.
Width (Metric) 4.2 m
Displayed Text BIOLOGICAL HAZARD AUTHORIZED PERSONNEL ONLY
View More Specs

Products 3
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19140981
View Documents Promotion Details
Brady™
126695
Polyester
Each for $36.41
Only null left
 
19140989
View Documents Promotion Details
Brady™
126694
Plastic
Each for $20.40
Only null left
 
19140995
View Documents Promotion Details
Brady™
126693
Aluminum
Each for $35.04
Only null left
 
Description

Description

  • Made from aluminum, plastic or self-sticking polyester
  • Size: 10 in. H x 14 in. W
  • Legend: BIOLOGICAL HAZARD AUTHORIZED PERSONNEL ONLY

Specifications

Specifications

10 in.
14 in.
BIOLOGICAL HAZARD AUTHORIZED PERSONNEL ONLY
3 m
4.2 m
Biohazard Identification Sign
Videos
SDS
Documents

Documents