missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Biohazard Identification Signs: Biological Hazard Authorized Personnel Only

For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents

Supplier:  Brady™ 126694

 View more versions of this product

Catalog No. 19140989


Only null left

Description

Description

  • Made from aluminum, plastic or self-sticking polyester
  • Size: 10 in. H x 14 in. W
  • Legend: BIOLOGICAL HAZARD AUTHORIZED PERSONNEL ONLY

Specifications

Chemical Identifiers

10 in.
14 in.
BIOLOGICAL HAZARD AUTHORIZED PERSONNEL ONLY
3 m
4.2 m
Biohazard Identification Sign

Specifications

Plastic
3 m
4.2 m
Biohazard Identification Sign
10 in.
14 in.
BIOLOGICAL HAZARD AUTHORIZED PERSONNEL ONLY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions