missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Biohazard Identification Signs: Biological Hazard Authorized Personnel Only
Description
- Made from aluminum, plastic or self-sticking polyester
- Size: 10 in. H x 14 in. W
- Legend: BIOLOGICAL HAZARD AUTHORIZED PERSONNEL ONLY
Specifications
Specifications
| Material | Aluminum |
| Mounting Type | Corner Holes |
| Height (English) | 10 in. |
| Height (Metric) | 3 m |
| Width (English) | 14 in. |
| Width (Metric) | 4.2 m |
| Shape | Rectangular |
| Languages | English |
| Displayed Text | BIOLOGICAL HAZARD AUTHORIZED PERSONNEL ONLY |
| Product Type | Biohazard Identification Sign |
| Show More |