missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Biohazard Identification Signs: Biological Hazard Authorized Personnel Only

Catalog No. 19140981
Change view
Click to view available options
Material:
Aluminum
Plastic
Polyester
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19140981 Polyester
19140989 Plastic
19140995 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19140981 Supplier Brady™ Supplier No. 126695

For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents

  • Made from aluminum, plastic or self-sticking polyester
  • Size: 10 in. H x 14 in. W
  • Legend: BIOLOGICAL HAZARD AUTHORIZED PERSONNEL ONLY

Specifications

Material Polyester
Description Self-sticking Polyester
Height (English) 10 in.
Height (Metric) 3 m
Width (English) 14 in.
Width (Metric) 4.2 m
Displayed Text BIOLOGICAL HAZARD AUTHORIZED PERSONNEL ONLY
Product Type Biohazard Identification Sign