missing translation for 'onlineSavingsMsg'
Learn More

Oakton™ Waterproof pH 450 Portable Meter

Numéro de catalogue. p-4954013
Change view
Click to view available options
Type:
pH Meter with Calibration Certificate and All-in-one pH/Temp. Electrode
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Type
15500120 pH Meter with Calibration Certificate and All-in-one pH/Temp. Electrode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 15500120 Fournisseur Oakton™ Code fournisseur 3561833
Il en reste null

Rugged design with backlit display for use in any environment

  • Intuitive, user-friendly icons show electrode status as pH slope
  • IP67 waterproof
  • Grip-Clip™ holder for quick and easy attachment of electrode to beaker or container
  • Large, backlit LCD displays measurement simultaneously with temperature and calibration data with % slope
  • Up to 6-point calibration from USA, NIST™ DIN and custom pH buffers
  • Automatic temperature compensation (ATC) for measurement accuracy, even in fluctuating temperatures
  • Perform data logging with manual or timed collection of up to 500 data sets with real-time clock time/date stamp
  • Download and analyze data sets with USB or RS-232 output
  • Measuring Range: -2.00 to 16.00 (pH); ±2000 (mV); -10° to +110°C (temp.); 0.0 to 19,999 (Ion)

  • Select temperature readout from °C/°F to suit your application
  • Other features include stability indicator, smart averaging, hold function, visible calibration alarm, battery life indicator, electrode status
  • Two AA batteries with extended battery life or optional universal power adapter
  • Built-in stand with wall mount option allows for easy benchtop measurement or storage

pH 450 Portable Meter Models

  • Cat. No. 15-500-119: Includes Grip-Clip probe holder and two AA batteries
  • Cat. No. 15-500-121: Includes NIST-Traceable calibration, Grip-Clip probe holder and two AA batteries
  • Cat. No. 15-500-118: Includes All-in-one pH/temperature electrode, Grip-Clip probe holder and two AA batteries
  • Cat. No. 15-500-120: Includes NIST™-Traceable calibration, All-in-one pH/temperature electrode, Grip-Clip probe holder and two AA batteries
  • Cat. No. 15-500-122: Includes All-in-one pH/temperature electrode, Grip-Clip probe holder, buffers and electrode storage solution, two AA batteries and hard carrying case
  • Cat. No. 15-500-123: Includes NIST-Traceable calibration, All-in-one pH/temperature electrode, Grip-Clip probe holder, buffers and electrode storage solution, two AA batteries and hard carrying case

Applications:

Ideal for manufacturing floor, field testing and laboratory

Certifications

CE marked

Warranty

Three years

Spécifications

Data Management Details Manual or Timed Collection
Accuracy (mV) ±0.2
Accuracy (Temperature) ±0.5°C
Accuracy (pH) ±0.01
Outputs USB, RS-232
Includes Calibration Certificate, All-in-one pH/temp electrode, Grip-Clip electrode holder and two AA batteries
For Use With (Application) Manufacturing floor, field testing and laboratory
Range (pH) -2 to 16
Resolution (pH) 0.01
Range (mV) ±2000
Resolution (mV) 0.1mV ±999.9mV; 1mV beyond
Calibration Points pH: Up to 6 (USA, NIST, DIN); ±200 mV; Up to 6 (Ion); 0.1° increments (temp.)
Temperature Range (Metric) -10°C to +110°C
Temperature Range (English) 17°F to 230°F
Resolution (Temperature) 0.1°
Display Type LCD, Graphic with Backlight
Data Points 500 Sets
Certifications/Compliance CE
IP Rating IP67
AC Adapter Optional universal AC adapter
Dimensions (L x W x H) 20 x 8 x 5.7 cm (7.87 x 3.12 x 2.25 in.)
Probe Type All-in-one pH/temperature electrode
Range (Ion) 0.0 to 19,999
Resolution (Ion) 0.01
Temperature Compensation Automatic or Manual
Battery Type 2 x AA
Type pH Meter with Calibration Certificate and All-in-one pH/Temp. Electrode
Battery Life 500 hr.
Length (English) 7.87 in.
Length (Metric) 20 cm
Width (English) 3.12 in.
Width (Metric) 8 cm
Height (English) 2.25 in.
Height (Metric) 5.7 cm
Accuracy (Ion) ±0.5% full-scale (monovalent); ±1% full-scale (divalent)
Afficher plus de résultats Afficher moins de résultats