missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Traceable™ Refrigerator/Freezer Ultra™ Thermometer
Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.
Supplier: Fisherbrand™ 9367806
Comprend: Flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips
Description
Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.
- Designed to meet current CDC requirements for vaccine storage and monitoring
- Triple display simultaneously shows min., max. and current temperatures for a specified probe
- Smart-Alarm™ with visual and audible indicators signal continuously when an out-of-range condition is sensed
- Alarm continues even after unit returns to non-alarm conditions and is programmable in 0.1° increments
- °C/°F switchable
- Enclosed temperature-buffered sensor to accurately monitor temperatures in one or two different areas in refrigerators, freezers, water baths and incubators
- Solid-state probe sealed in a miniature bottle (1 x 2.5 in.) eliminates mercury contamination and rapid temperature changes when refrigerator door is opened
- Miniature bottle is filled with nontoxic glycol, generally recognized as safe (GRAS) by FDA thereby eliminating concerns about contact with food or drinking water
- Features high-impact, chemical-resistant ABS plastic case (2.75 x 4.25 x 0.75 in.)
- Ten-foot micro-cable permits refrigerator doors to close on it
- Range: -50° to +70°C (-58° to +158°F)
- Accuracy: ±0.5°C between -30° to +50°C, otherwise ±1°C
- Resolution: 0.1°
- Sampling Rate: 30 seconds
Certifications
Individually serial-numbered Traceable™ Certificate is provided from ISO/IEC 17025:2005 calibration laboratory accredited by A2LA; indicates traceability to standards provided by NIST.
Identifiants chimiques
| 2.5 x 6.3 cm (1 x 2.5 in.) | |
| 0.1° | |
| Refrigerator/Freezer Ultra Thermometer | |
| 70°C | |
| -50°C | |
| -50°C to +70°C | |
| Monitoring Temperature in Freezer, Water Bath, Heating Block, Incubator and Refrigerator | |
| Digital |
| Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators | |
| 4 oz. | |
| 158°F | |
| -58°F | |
| -58°F to +158°F | |
| Yes | |
| ±0.5°C (-30°C to +50°C), ±1°C otherwise | |
| 6.99 cm |
Spécifications
| Nontoxic Glycol | |
| NIST Traceable | |
| Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators | |
| 7 cm | |
| 1 x 5 mL Vaccine Bottle Probe | |
| 4 oz. | |
| Refrigerator/Freezer Ultra Thermometer | |
| 158°F | |
| -58°F | |
| -58°F to +158°F | |
| Yes | |
| ±0.5°C (-30°C to +50°C), ±1°C otherwise | |
| 6.99 cm |
| 2.5 x 6.3 cm (1 x 2.5 in.) | |
| 7 x 10.8 x 2 cm (2.75 x 4.25 x 0.75 in.) | |
| Traceable™ Certificate, flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips | |
| ABS Plastic | |
| 0.1° | |
| 113.4 g | |
| 3 in. | |
| 70°C | |
| -50°C | |
| -50°C to +70°C | |
| Monitoring Temperature in Freezer, Water Bath, Heating Block, Incubator and Refrigerator | |
| Digital |