missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. V00751G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V00751G Supplier TCI America Supplier No. V00751G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Vinylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 2156-04-9
Color White
Molecular Formula C8H9BO2
Quantity 1 g
MDL Number MFCD00239441
Synonym 4-vinylphenylboronic acid, 4-vinylbenzeneboronic acid, 4-vinylphenyl boronic acid, 4-ethenylphenyl boronic acid, 4-vinylboronic acid, p-styrylboronic acid, p-vinylphenylboronic acid, styrene-4-boronic acid, boronic acid, 4-ethenylphenyl, 4-styreneboronic acid
InChI Key QWMJEUJXWVZSAG-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(C=C)C=C1
IUPAC Name (4-ethenylphenyl)boronic acid
Molecular Weight (g/mol) 147.97
PubChem CID 2734393
Formula Weight 147.97
Physical Form Crystalline Powder
Show More Show Less