missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzyl acetate, 95%, stabilized, Thermo Scientific™

Numéro de catalogue. p-3779890
Modifier l'affichage
Click to view available options
Quantité:
5g
25g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC422500050 5g Glass bottle
AC422500250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC422500050 Fournisseur Thermo Scientific Chemicals Code fournisseur 422500050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 1592-12-7
Formule moléculaire C11H12O2
Poids moléculaire (g/mol) 176.22
Numéro MDL MFCD00078265
Clé InChI LEIKPUSDAWATBV-UHFFFAOYSA-N
Synonyme 4-vinylbenzyl acetate, 4-ethenylphenyl methyl acetate, 4-acetoxymethylstyrene, 4-vinylbenzylacetate, p-acetoxymethylstyrene, 4-vinylphenyl methyl acetate, 4-ethenylbenzenemethanol acetate, benzenemethanol,4-ethenyl-, 1-acetate
PubChem CID 2735161
Nom de l’IUPAC (4-ethenylphenyl)methyl acetate
SOURIRES CC(=O)OCC1=CC=C(C=C)C=C1

Spécifications

Nom ou substance chimique 4-Vinylbenzyl acetate
Nom Remarque Stabilized
CAS % minimum 94.0
CAS % maximal 100.0
Densité 1.5300g/mL
Point d’ébullition 125°C (12.0mmHg)
Point d’éclair >93°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 94% min. (GC)
Emballage Glass bottle
Indice de réfraction 1.5350 to 1.538
Quantité 5g
Gravité spécifique 1.53
Stabilisateur 0.1 % hydroquinone
Poids de formule 176.22
Pourcentage de pureté 95%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only