missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethylbenzoic Anhydride 97.0+%, TCI America™

Catalog No. T159310G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T159310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T159310G Supplier TCI America Supplier No. T159310G
Only null left

Chemical Identifiers

CAS 25753-16-6
Molecular Formula C16H8F6O3
Molecular Weight (g/mol) 362.227
MDL Number MFCD00671577
InChI Key FNAWJOBKLWLHTA-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoic anhydride, tfba, 4-trifluoromethylbenzoic anhydride, 4-trifluoromethyl benzoyl 4-trifluoromethyl benzoate, benzoic acid,4-trifluoromethyl-, anhydride with 4-trifluoromethyl benzoic acid, acmc-1cg53, p-trifluoromethylbenzoic acid anhydride, bis 4-trifluoromethylbenzoic acid anhydride, benzoic acid, 4-trifluoromethyl-, anhydride
PubChem CID 2760733
IUPAC Name [4-(trifluoromethyl)benzoyl] 4-(trifluoromethyl)benzoate
SMILES C1=CC(=CC=C1C(=O)OC(=O)C2=CC=C(C=C2)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 130°C
Color White
Quantity 10 g
Formula Weight 362.23
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Trifluoromethylbenzoic Anhydride