missing translation for 'onlineSavingsMsg'
Learn More

[(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate 97.0+%, TCI America™

Catalog No. T3622200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3622200MG 200 mg
T36221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3622200MG Supplier TCI America Supplier No. T3622200MG

Chemical Identifiers

CAS 1868173-15-2
Molecular Formula C23H22F3IO6S
Molecular Weight (g/mol) 610.38
MDL Number MFCD30725767
InChI Key RTMJODUYHFSMBJ-UHFFFAOYSA-M
Synonym [(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium Tosylate
PubChem CID 131697921
IUPAC Name [4-(trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodanium 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S([O-])(=O)=O.COC1=CC(OC)=C([I+]C2=CC=C(C=C2)C(F)(F)F)C(OC)=C1

Specifications

Melting Point 171°C
Quantity 200 mg
Formula Weight 610.38
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [(4-Trifluoromethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator