missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzylamine 98.0+%, TCI America™
Supplier: TCI America T150825G
Specifications
| 4-(Trifluoromethyl)benzylamine | |
| Yellow | |
| C8H8F3N | |
| 25 g | |
| 4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl | |
| NCC1=CC=C(C=C1)C(F)(F)F | |
| 175.15 | |
| 175.15 | |
| Liquid |
| 3300-51-4 | |
| 113°C | |
| MFCD00010220 | |
| 1760 | |
| PRDBLLIPPDOICK-UHFFFAOYSA-N | |
| 1-[4-(trifluoromethyl)phenyl]methanamine | |
| 76804 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 3300-51-4 | |
| 175.15 | |
| PRDBLLIPPDOICK-UHFFFAOYSA-N | |
| 76804 | |
| NCC1=CC=C(C=C1)C(F)(F)F |
| C8H8F3N | |
| MFCD00010220 | |
| 4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl | |
| 1-[4-(trifluoromethyl)phenyl]methanamine |
Safety and Handling
TSCA : No