missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)benzylamine 98.0+%, TCI America™
$83.89 - $329.94
Chemical Identifiers
| CAS | 3300-51-4 |
|---|---|
| Molecular Formula | C8H8F3N |
| Molecular Weight (g/mol) | 175.15 |
| MDL Number | MFCD00010220 |
| InChI Key | PRDBLLIPPDOICK-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl |
| PubChem CID | 76804 |
| IUPAC Name | 1-[4-(trifluoromethyl)phenyl]methanamine |
| SMILES | NCC1=CC=C(C=C1)C(F)(F)F |
Chemical Identifiers
| 3300-51-4 | |
| 175.15 | |
| PRDBLLIPPDOICK-UHFFFAOYSA-N | |
| 76804 | |
| NCC1=CC=C(C=C1)C(F)(F)F |
| C8H8F3N | |
| MFCD00010220 | |
| 4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl | |
| 1-[4-(trifluoromethyl)phenyl]methanamine |
Specifications
| 3300-51-4 | |
| 113°C | |
| MFCD00010220 | |
| 1760 | |
| PRDBLLIPPDOICK-UHFFFAOYSA-N | |
| 1-[4-(trifluoromethyl)phenyl]methanamine | |
| 76804 | |
| ≥98.0% (GC,T) | |
| 4-(Trifluoromethyl)benzylamine |
| Yellow | |
| C8H8F3N | |
| 5 g | |
| 4-trifluoromethyl benzylamine, 4-trifluoromethylbenzylamine, 4-trifluoromethyl phenyl methanamine, 1-4-trifluoromethyl phenyl methanamine, 4-trifluoromethyl benzyl amine, p-trifluoromethylbenzyl amine, 4-trifluoromethyl-benzylamine, p-trifluoromethyl benzylamine, benzenemethanamine, 4-trifluoromethyl | |
| NCC1=CC=C(C=C1)C(F)(F)F | |
| 175.15 | |
| 175.15 | |
| Liquid |
Safety and Handling
TSCA : No