accessibility menu, dialog, popup

UserName

CAS RN 3300-51-4

IUPAC Name: 1-[4-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[4-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 175.15
Molecular Formula: C8H8F3N
InChi Key: PRDBLLIPPDOICK-UHFFFAOYSA-N
SMILES: NCC1=CC=C(C=C1)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)